Agarose 4% B - CL, cross-linked, beads

Supplier: Thermo Fisher Scientific
Warning

ALFAH32305.AUEA 0 SGD
ALFAH32305.AU
Agarose 4% B - CL, cross-linked, beads
Agarose

CAS No.: 61970-08-9 Storage Temperature: 2°C to 8°C Flash Point: 38°(100°F) Boiling Point: 100° UN No.: UN1170 MDL No.: MFCD00081294

Boiling Pt: 100 °C
Density: 1,040
Flash Pt: 38 °C (100 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00081294
CAS Number: 9012-36-6
UN: 1170
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR