Volumetric flasks, class A, blue markings
Supplier: SciLabware
FRD178
BIBBFRD178EA 129 SGD
BIBBFRD178
Volumetric flasks, class A, blue markings
Flasks Volumetric Flasks
MBL®, borosilicate glass, with PE stopper.
  • Marks and inscriptions in high contrast blue enamel, conformity 'H' mark
  • Each flask is laser etched with a unique individual serial number, batch code and date of manufacture - Batch certificates are downloadable from website: www.scilabware.com
  • Calibrated to contain (TC, In)
Certifications: DIN 12664, BS EN ISO 1042