Centrifuge tubes conical bottom, Corning®
Supplier: Corning
11750
CORN11750EA 571 SGD
CORN11750
Centrifuge tubes conical bottom, Corning®
Tubes Centrifuge Tubes
PP sterile closed system centrifuge tubes arrive individually double bagged and pre-assembled. The caps have two ports: One port ends in a 0,2 µm filter, and the other port is chemically resistant, heat sealable flexible tubing, ending in a male Luer. Dip tubes, where available, reach all the way to the bottom of the flask for easy aseptic transfer during your liquid handling process.
These centrifuge tubes are ideal for applications requiring large volume centrifugation.
Each case of tubes contains a recyclable foam rack to facilitate handling. Support cushions must be used with thisMore Product Information