Culture flasks, low form, PYREX®
Supplier: Corning
4422-2XL
CORN4422-2XLEA 302 SGD
CORN4422-2XL
Culture flasks, low form, PYREX®
Flasks Culture Flasks
PYREX® borosilicate glass.
  • Very good temperature resistance
  • Very good chemical resistance
These low form culture flasks are ideal for culturing organisms that require a large surface area-to-volume ratio, and in serum production. These flask have a heavy beaded rim to protect against chipping and include a large marking spot allows for accurate labeling and easy identification.
Caution: Do not use this flask on a heat source smaller than the base of the flask. Uneven heating of the flask could cause breakage.