Hydrochloric acid 1% Reag. Ph. Eur.

Supplier: VWR Chemicals
Warning

Synonyms: Hydrochloric acid solution, Muriatic acid

VWRC87684.290EA 272 SGD
VWRC87684.290
Hydrochloric acid 1% Reag. Ph. Eur.
Hydrochloric acid
Formula: HCl
Boiling Pt: >100 °C (1013 hPa)
Density: 1,02…1.05 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR