VWR® Fixing Solution
Supplier: VWR Chemicals
VWRC87969.260EA 188 SGD
VWRC87969.260
VWR® Fixing Solution
Stains Microscopy and staining reagents
Fixing solution with 0,02% formaldehyde and 0,01% citric acid in aqueous solution.
Ordering information: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.