VWR® Q path® Safesolv, Solvent Substitute
Supplier: VWR Q-Path Chemicals
VWRQ00699464EA 64.6 SGD
VWRQ00699464
VWR® Q path® Safesolv, Solvent Substitute
Stains Microscopy and staining reagents
Q Path® Safesolv is a non hazardous replacement for xylene or toluene. It is compatible with the Q Path® Safemount mounting medium reference 00647520. It is used in place of xylene, toluene, methylecyclohexane or other solvents in the course of the dehydration stages of histological specimens and during the stages of staining and lamella mounting.
  • Clear colourless liquid