VWR®, Cleanroom Hoods, Irradiated
Supplier: VWR Collection
414004-502
VWRU414004-502EA 419 SGD
VWRU414004-502
VWR®, Cleanroom Hoods, Irradiated
Disposable Garments
This irradiated hood is manufactured from a specially formulated breathable microporous fabric that provides significantly fluid and barrier protection.
  • Excellent water vapour transmission to optimise user comfort
  • Hood is gamma irradiated to a Sterility Assurance Level (SAL) of 10⁻⁴ to address the concern of bio-burden contamination
  • Lowest level of particle shedding
  • Durable yet soft, cloth-like fabric
Packaging: Packed individually in sealed polyethylene bags contained within a larger carton liner. Each case is supplied with a certificate of irradiation.