4-Chloro-8-methoxy-2-methylquinoline 95%

Supplier: Thermo Scientific
Warning

H50559.03 H50559.06
ALFAH50559.03EA 0 SGD
ALFAH50559.03 ALFAH50559.06
4-Chloro-8-methoxy-2-methylquinoline 95%
4-Chloro-8-methoxy-2-methylquinoline

Purity: 95% CAS No.: 64951-58-2 Molecular Formula: C11H10ClNO Formula Weight: 207.66 Melting Point: 89-93° MDL No.: MFCD00272401

Formula: C₁₁H₁₀ClNO
MW: 207,66 g/mol
Melting Pt: 89…93 °C
Storage Temperature: Ambient
MDL Number: MFCD00272401
CAS Number: 64951-58-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR