6-Methoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride 95%
Supplier: Thermo Scientific
|
Warning
|
H31910.MD
H31910.03
ALFAH31910.MDEA
0
SGD
ALFAH31910.MD
ALFAH31910.03
6-Methoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride 95%
6-Methoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride
Purity: 95%
CAS No.: 57196-62-0
Molecular Formula: C10H13NO.HCl
Formula Weight: 199.68
Melting Point: 233-236°
MDL No.: MFCD01717328
Formula:
C₁₀H₁₄ClNO MW: 199,68 g/mol Melting Pt: 233…236 °C Storage Temperature: Ambient |
MDL Number:
MFCD01717328 CAS Number: 57196-62-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...