(S)-1,2,3,4-Tetrahydroisoquinoline-3-acetic acid hydrochloride 95%

Supplier: Thermo Scientific

H52111.MD H52111.03
ALFAH52111.MDEA 454 SGD
ALFAH52111.MD ALFAH52111.03
(S)-1,2,3,4-Tetrahydroisoquinoline-3-acetic acid hydrochloride 95%
(S)-1,2,3,4-Tetrahydroisoquinoline-3-acetic acid hydrochloride

Purity: 95% CAS No.: 270082-22-9 Molecular Formula: C11H13NO2.HCl Formula Weight: 227.69 MDL No.: MFCD01861030

Formula: C₁₁H₁₃NO₂·HCl
MW: 227,69 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01861030
CAS Number: 270082-22-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR