HYCARE Hand Sanitizer
Supplier: ELITE ADVANCED MATERIALS SDN
HYC111-0.1P HYC111-0.1P-LQ HYC111-0.5P HYC111-0.5P-LQ HYC111-1.0P HYC111-1.0P-LQ HYC111-2.5P HYC111-2.5P-LQ HYC111-25P HYC111-25P-LQ HYC111-5.0P HYC111-5.0P-LQ
ELITHYC111-0.1PEA 2.3 SGD
ELITHYC111-0.1P ELITHYC111-0.1P-LQ ELITHYC111-0.5P ELITHYC111-0.5P-LQ ELITHYC111-1.0P ELITHYC111-1.0P-LQ ELITHYC111-2.5P ELITHYC111-2.5P-LQ ELITHYC111-25P ELITHYC111-25P-LQ ELITHYC111-5.0P ELITHYC111-5.0P-LQ
HYCARE Hand Sanitizer
Cleaning Agents Disinfectants
HYCARE Hand sanitizer is an alcohol-based handrub with WHO-recommended formulation (75% Isopropyl alcohol).
  • Evidence-based, intrinsic advantages of fast acting and broad-spectrum microbiocidal activity with a minimal risk of generating resistance to antimicrobial agents
  • Suitability for use in resource-limited or remote areas with lack of accessibility to sinks or other facilities for hand hygiene (including clean water, towels)
  • Capacity to promote improved compliance with hand hygiene by making the process faster and more convenient
  • Economic benefit by reducing annual costs for hand hygiene
  • Minimization of risks from adverse events because of increased safety associated with better acceptability and tolerance than other products
The microbicidal activity of the WHO-recommended formulations was tested by WHO reference laboratories according to EN standards (EN 1500). Fragrance-free to reduce the risk of allergic reactions. Strict quality control and testing in qualified laboratory to ensure composition is met to standard and no harmful chemicals like methanol are present. Economic yet effective.