N-Methyl-2-pyrrolidone (NMP) ≥99.5%, B&J Brand™, Burdick & Jackson™

Supplier: BURDICK JACKSON
Danger

304-1 304-4 NP304-20
BJAA304-1EA 0 SGD
BJAA304-1 BJAA304-4 BJAANP304-20
N-Methyl-2-pyrrolidone (NMP) ≥99.5%, B&J Brand™, Burdick & Jackson™
N-Methyl-2-pyrrolidone (NMP)

  • Minimal UV contaminants
  • High GC purity
  • Low water content
  • Outstanding lot-to-lot consistency

Formula: C₅H₉NO
Boiling Pt: 202 °C
Melting Pt: -24 °C
Density: 1,0259 g/cm³ (25 °C)
Flash Pt: 9,5 %(V)
Storage Temperature: Ambient
MDL Number: MFCD00003193
CAS Number: 872-50-4
EINECS: 212-828-1
Merck Index: 13,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32d156c242c81ef2974339109ced3895beda87fd&vid=06c87bbb3b5050362f47a8fc836cedc8a6d51530&prid=web&clid=SAPDAM

Order Now

Specification Test Results

GC Analysis 99.5 % min.
Refractive Index (20ºC) 1.4680 - 1.4720
UV Absorbance @ 300 nm 0.5 AU max.
UV Absorbance @ 325 nm 0.1 AU max.
UV Absorbance @ 350 nm 0.03 AU max.
UV Absorbance @ 400 nm 0.01 AU max.
UV Cutoff 285 nm max.
Water by Karl Fischer Titration 0.05 % max.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR