Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: ABCAM
AB6789-1
ABCAAB6789-1EA 0 SGD
ABCAAB6789-1
Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Type: Secondary
Antigen: IgG (Mouse) (mouse IgG)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

SPECIFICATIONS

Antigen symbol IgG
Environmentally Preferable
Antibody type Secondary
Clonality Polyclonal
Conjugation HRP (Horseradish Peroxidase)
Reactivity
Host Goat
Isotype
Western blot Yes
ELISA Yes
ImmunoChemistry Yes
ImmunoPrecipitation Yes
ImmunoFluorescence Yes
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR