Anti-SDCBP Mouse Monoclonal Antibody [clone: 2C12]

Supplier: Abnova
H00006386-M01
ABNOH00006386-M01EA 0 SGD
ABNOH00006386-M01
Anti-SDCBP Mouse Monoclonal Antibody [clone: 2C12]
Antibodies
Type: Primary
Antigen: SDCBP
Clonality: Monoclonal
Clone: 2C12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol SDCBP
Environmentally Preferable
Antigen name Syndecan Binding Protein
Antigen synonyms syndecan binding protein (syntenin)|ST1|TACIP18 |SYCL|pro-TGF-alpha cytoplasmic domain-interacting protein 18|MDA-9|melanoma differentiation associated protein-9|SDCBP|scaffold protein Pbp1 syntenin|Syntenin
Antibody type Primary
Clonality Monoclonal
Conjugation Unconjugated
Clone 2C12
Reactivity Human
Host Mouse
Gene ID 6386
Isotype
Western blot Yes
ELISA Yes
ImmunoChemistry Yes
ImmunoPrecipitation Yes
ImmunoFluorescence Yes
Cross adsorption No
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR