Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Abnova
PAB0096
ABNOPAB0096EA 0 SGD
ABNOPAB0096
Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Order Now

SPECIFICATIONS

Antigen symbol IgG
Environmentally Preferable
Antigen name Immunoglobulin G
Antibody type Secondary
Clonality Polyclonal
Conjugation HRP (Horseradish Peroxidase)
Reactivity Mouse
Host Goat
Isotype
Western blot Yes
ELISA Yes
ImmunoChemistry Yes
Cross adsorption No
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR