Ace-Thred Reducing coupling
Supplier: Ace Glass
5842-46 5842-48
ACEG5842-46EA 0 SGD
ACEG5842-46 ACEG5842-48
Ace-Thred Reducing coupling
Fittings and Connectors
These couplings are designed for threading columns together, or to secure end fittings with a single O-ring seal for leak-tight engagement, with no significant size reduction in the Int.Ø.
One end is threaded for the next smallest diameter. Size listed refers to inside diameter of threads and corresponds to the column diameter. Fabricated from borosilicate glass.