Stirring Shaft, Borosilicate
Supplier: Ace Glass
8075-12
ACEG8075-12EA 545 SGD
ACEG8075-12
Stirring Shaft, Borosilicate
Stirrers Stirring Shafts
Polished glass stirring shafts, External diameter of 10 mm.
Can be used to join blades to button type shafts, and join agitators to beaded shafts. Plain shafts will accept agitators secured with a set screw anywhere along the ground flat section. Can also be joined to stirrer motor with the use of a chuck. Fabricated from borosilicate glass.