Pressure Tubes with Ace-Thred
Supplier: Ace Glass
8648-30
ACEG8648-30EA 0 SGD
ACEG8648-30
Pressure Tubes with Ace-Thred
Reaction Vessels Reaction Vessels, Cylindrical Shape
Heavy-wall, borosilicate glass tubes are designed for polymerisation and analysis of halogens by thermal decomposition in the presence of lime
They feature the unique Ace-Thred closure system for assurance of a secure seal. Tubes maintain a recommended operating pressure of 150 psi. Hydrogenation bottles also feature the Ace-Thred closure system and are flat bottom vessels. These bottles maintain a lower recommended operating pressure, not to exceed 60psi. Tubes and bottles are supplied without plugs and O-rings. Tube length is measured below the thread. Tube pressure ratings areMore Product Information