Avantor® ACE® SuperC18, HPLC Columns, 5 µm

Supplier: Avantor
ACETACE-1211-1010EA 0 SGD
ACETACE-1211-1010 ACETACE-1211-1020 ACETACE-1211-1220 ACETACE-1211-1510 ACETACE-1211-1520 ACETACE-1211-2510 ACETACE-1211-2520 ACETACE-1211-0510 ACETACE-1211-0520 ACETACE-1211-2530 ACETACE-1211-15005 ACETACE-12110103GD ACETACE-12110102GD ACETACE12110110GD ACETACE12110120GD ACETACE-1211-0530 ACETACE-1211-7530 ACETACE-1211-1030 ACETACE-1211-1530 ACETACE-1211-3030
Avantor® ACE® SuperC18, HPLC Columns, 5 µm
Chromatography Columns
Avantor® ACE® SuperC18 columns provide high stability across an extended pH range. These stainless steel columns are ideal for use with LC/MS compatible buffers and can be used with both MeOH and MeCN mobile phases.

  • Ultra-inert for maximum performance and reproducibility
  • Ultra low bleed for improved LC/MS compatibility
  • Wide range of particle sizes (1,7 to 10 µm) and complementary phases

Maximum recommended pH: 1,5 to 11,5

Max. pressure: 275 bar
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR