Avantor® ACE® C4, HPLC Columns, 5 µm
Supplier: Avantor
ACETACE-123-0246EA 0 SGD
ACETACE-123-0246 ACETACE-123-0302 ACETACE-123-0502 ACETACE-123-0520 ACETACE-123-0530 ACETACE-123-0546 ACETACE-123-1010 ACETACE-123-1020 ACETACE-123-1030 ACETACE-123-1204 ACETACE-123-1220 ACETACE-123-1510 ACETACE-123-1520 ACETACE-123-1530 ACETACE-123-1546 ACETACE-123-2502 ACETACE-123-2504 ACETACE-123-2510 ACETACE-123-2520 ACETACE-123-2530 ACETACE-123-2546 ACETACE-123-3503 ACETACE-123-7502 ACETACE-123-7520 ACETACE-123-0103GD ACETACE-123-0110GD ACETACE-123-7530 ACETACE-123-3020 ACETACE-123-3030 ACETACE-123-10003 ACETACE-123-25003 ACETACE-123-03003 ACETACE-123-1539
Avantor® ACE® C4, HPLC Columns, 5 µm
Chromatography Columns
Avantor® ACE® C4 columns provide great reproducibility and column lifetime. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative.
  • Ultra high purity, base deactivated silica with exceptional reproducibility
  • Ideal for a wide range of analytes differing in hydrophobicity
  • Wide range of particle sizes (2 to 10 µm) and complementary phases
  • Method validation kits contain three different batches of the same phase - available on request
These columns are designed for a wide range ofMore Product Information