Avantor® ACE® C8, HPLC Columns, 10 µm
Supplier: Avantor
ACETACE-132-0510EA 0 SGD
ACETACE-132-0510 ACETACE-132-0520 ACETACE-132-0530 ACETACE-132-1010 ACETACE-132-1020 ACETACE-132-1030 ACETACE-132-1046 ACETACE-132-1220 ACETACE-132-1510 ACETACE-132-1520 ACETACE-132-1530 ACETACE-132-1546 ACETACE-132-2510 ACETACE-132-2520 ACETACE-132-2530 ACETACE-132-2546 ACETACE-132-7520 ACETACE-132-0110GD ACETACE-132-7530 ACETACE-132-3020 ACETACE-132-3030 ACETACE-132-3046 ACETACE-132-2504
Avantor® ACE® C8, HPLC Columns, 10 µm
Chromatography Columns
These ultra-inert Avantor® ACE® C8 columns provide great reproducibility and column lifetime. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative.
  • Ultra high purity, base deactivated silica with exceptional reproducibility
  • Ideal for a wide range of analytes differing in hydrophobicity
  • Wide range of particle sizes (2 µm to 10 µm) and complementary phases
  • Can be used as general purpose starting point column
  • Method validation kits contain three different batches of the same phase -
More Product Information