Avantor® ACE® CN, HPLC Columns, 10 µm
Supplier: Avantor
ACETACE-134-0510EA 0 SGD
ACETACE-134-0510 ACETACE-134-0520 ACETACE-134-0530 ACETACE-134-1010 ACETACE-134-1020 ACETACE-134-1030 ACETACE-134-1220 ACETACE-134-1246 ACETACE-134-1510 ACETACE-134-1520 ACETACE-134-1530 ACETACE-134-2510 ACETACE-134-2520 ACETACE-134-2530 ACETACE-134-2546 ACETACE-134-7520 ACETACE-134-0110GD ACETACE-134-7530 ACETACE-134-3020 ACETACE-134-3030 ACETACE-134-2504 ACETACE-134-3004 ACETACE-134-1539
Avantor® ACE® CN, HPLC Columns, 10 µm
Chromatography Columns
These Avantor® ACE® CN columns provide great reproducibility and column lifetime.
  • Ultra high purity, base deactivated silica with exceptional reproducibility
  • Can be used under both reversed phase and normal phase conditions
  • Capillary to preparative dimensions
  • Wide range of particle sizes (2 µm to 10 µm) and complementary phases
  • Useful for the separation of polar analytes and compounds with double and triple bonds
  • Method validation kits contain three different batches of the same phase - available on request
These stainless steel columns are availableMore Product Information