Avantor® ACE® AQ, HPLC Columns, 10 µm
Supplier: Avantor
ACETACE-136-0510EA 0 SGD
ACETACE-136-0510 ACETACE-136-0520 ACETACE-136-0530 ACETACE-136-1010 ACETACE-136-1020 ACETACE-136-1030 ACETACE-136-1220 ACETACE-136-1510 ACETACE-136-1520 ACETACE-136-1530 ACETACE-136-2510 ACETACE-136-2520 ACETACE-136-2530 ACETACE-136-7520 ACETACE-136-0103GD ACETACE-136-0110GD ACETACE-136-7530 ACETACE-136-3020 ACETACE-136-3030 ACETACE-136-3004
Avantor® ACE® AQ, HPLC Columns, 10 µm
Chromatography Columns
Avantor® ACE® AQ columns provide great reproducibility and column lifetime. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative.
  • Ultra high purity, base deactivated silica with exceptional reproducibility
  • Retains polar compounds in 100% aqueous mobile phase
  • Unique C18 bonded phase with integral polar functionality
  • Ideal for use with fast gradients due to its rapid re-equilibration properties and resistance to retention loss
  • Wide range of particle sizes (2 µm to 10 µm) and complementary
More Product Information