Avantor® ACE®, Preparative Column Coupler
Supplier: Avantor
ACETC0002EA 0 SGD
ACETC0002
Avantor® ACE®, Preparative Column Coupler
Fittings and Connectors
Column coupler made from stainless steel/PEEK suitable for connecting preparative columns to stand-alone guard cartridge holder, or for making similar HPLC connections.
Ideally suited to connect 30 mm id preparative columns to H0009 cartridge holder.