Avantor® ACE® Excel® CN, HPLC/UHPLC Columns, Analytical, 2 µm
Supplier: Avantor
ACETEXL-104-1503UEA 0 SGD
ACETEXL-104-1503U ACETEXL-104-0502U ACETACE-1P4-GD2U ACETACE-1P4-GD4U
Avantor® ACE® Excel® CN, HPLC/UHPLC Columns, Analytical, 2 µm
Chromatography Columns
These Avantor® ACE® CN columns provide great reproducibility and column lifetime. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative.
  • Ultra high purity, base deactivated silica with exceptional reproducibility
  • Can be used under both reversed phase and normal phase conditions
  • Capillary to preparative dimensions
  • Wide range of particle sizes (2, 3 and 5 µm)
  • Useful for the separation of polar analytes and compounds with double and triple bonds
  • Method validation kits contain three different
More Product Information