Avantor® ACE® Excel® C4, HPLC/UHPLC Columns, Analytical, 5 µm
Supplier: Avantor
ACETEXL-123-1046UEA 0 SGD
ACETEXL-123-1046U ACETEXL-123-1546U ACETEXL-123-0202U ACETEXL-123-0203U ACETEXL-123-2546U ACETEXL-123-0502U ACETEXL-123-7502U
Avantor® ACE® Excel® C4, HPLC/UHPLC Columns, Analytical, 5 µm
Chromatography Columns
These ultra-inert Avantor® ACE® Excel® C4 columns provide great reproducibility and column lifetime. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative.
  • Ultra high purity, base deactivated silica with exceptional reproducibility
  • Ideal for a wide range of analytes differing in hydrophobicity
  • Wide range of particle sizes (2 to 5 µm) and complementary phases available
  • Method validation kits contain three different batches of the same phase - available on request
These columns areMore Product Information