Avantor® ACE® Excel® Phenyl, HPLC/UHPLC Columns, Analytical, 5 µm
Supplier: Avantor
ACETEXL-125-1002UEA 0 SGD
ACETEXL-125-1002U ACETEXL-125-1003U ACETEXL-125-1046U ACETEXL-125-1502U ACETEXL-125-1546U ACETEXL-125-2546U ACETEXL-125-0302U ACETEXL-125-0502U
Avantor® ACE® Excel® Phenyl, HPLC/UHPLC Columns, Analytical, 5 µm
Chromatography Columns
These Avantor® ACE® Excel® Phenyl columns provide great reproducibility and column lifetime. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative.
  • Ultra high purity, base deactivated silica with exceptional reproducibility
  • Offers strong π-π interactions
  • Ideal for analytes with π-bonding, electron withdrawing or delocalised groups
  • Wide range of particle sizes available (2 µm, 3 µm and 5 µm) and complementary phases
  • Method validation kits contain three different batches of the same phase -
More Product Information