VWR® ACE® Excel® C18-HL HPLC/UHPLC Columns, Analytical, 3 µm
Supplier: Avantor
ACETEXL-311-1546UEA 0 SGD
ACETEXL-311-1546U ACETEXL-311-2503U ACETEXL-311-2546U ACETEXL-311-2504U
VWR® ACE® Excel® C18-HL HPLC/UHPLC Columns, Analytical, 3 µm
Chromatography Columns
Avantor® ACE® Excel® C18-HL columns provide great reproducibility and column lifetime. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative.
  • Ultra high purity, base deactivated silica with exceptional reproducibility
  • Ideal for a wide range of analytes differing in hydrophobicity
  • Wide range of particle sizes (3 and 5 µm) and complementary phases
  • Especially suited for preparative work due to higher carbon load
  • Method validation kits contain three different batches of the same phase -
More Product Information