Anthranilamide 99+%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 2-Aminobenzamide

104901000
ACRO104901000EA 0 SGD
ACRO104901000
Anthranilamide 99+%
Anthranilamide
Formula: C₇H₈N₂O
Boiling Pt: 300 °C (760 mmHg)
Melting Pt: 111…114 °C (dec.)
Flash Pt: 198 °C
Storage Temperature: Ambient
MDL Number: MFCD00007981
CAS Number: 88-68-6
EINECS: 201-851-2

Order Now

Specification Test Results

Appearance Light brown to grey-brown Crystalline powder or flakes
Infrared spectrum Conforms
Melting point 111 to 114 °C
HPLC ≥99.0 %
Residue after evaporation ≤0.0005 %
Free amine ≤0.0002 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR