Molybdic acid for analysis

Supplier: Thermo Fisher Scientific
Warning

213621000
ACRO213621000EA 0 SGD
ACRO213621000
Molybdic acid for analysis
Molybdic acid
Formula: H₂MoO₄
Boiling Pt: ∼300 °C (1013 hPa)
Melting Pt: 300 °C
Density: 3,1 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00036287
CAS Number: 7782-91-4
EINECS: 231-970-5
Merck Index: 13,06262

Order Now

Specification Test Results

Appearance White to light yellow Powder
Additional info contains ammonium molybdate
Titration with NaOH ≥85 % (as MoO3)
Chloride (Cl) ≤100 ppm
Phosphate (PO4) ≤5 ppm
Sulfate (SO4) ≤2000 ppm
Iron (Fe) ≤50 ppm
Heavy metals (as Pb) ≤30 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR