L-Methionine sulphoximine 98+%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: (2S)-2-Amino-4-(S-methylsulphonimidoyl)butanoic acid, H-Met(sulphoximine)-OH

227200010
ACRO227200010EA 0 SGD
ACRO227200010
L-Methionine sulphoximine 98+%
L-Methionine sulphoximine

CAS No.: 15985-39-4

Formula: C₅H₁₂N₂O₃S
Melting Pt: 210 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002621
CAS Number: 15985-39-4

Order Now

Specification Test Results

Appearance White to almost white Crystalline powder
Infrared spectrum Conforms
Specific optical rotation +12° to +15° (20°C, 589 nm) (c=1, H2O)
Titration with HClO4 98.0 to 102.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR