Petroleum spirit, 100…140 °C, extra pure

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Gasoline, Petroleum ether, Petrol, Benzine

233020025
ACRO233020025EA 0 SGD
ACRO233020025
Petroleum spirit, 100…140 °C, extra pure
Petroleum spirit
Boiling Pt: 100…140 °C (760 mmHg)
Melting Pt: –30 °C
Density: 0,74 g/cm³
Flash Pt: 2 °C
Storage Temperature: Ambient
MDL Number: MFCD00081849
CAS Number: 64742-49-0
EINECS: 265-151-9
UN: 3295
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4070 to 1.4150 (20°C, 589 nm)
Boiling point 98 to 140°C
Density (15°C) 0.737 to 0.747 g/ml
Color scale ≤10 APHA
Residue after evaporation ≤0.005 %
Benzene ≤50 ppm
Aromatics ≤0.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR