Hyaluronic acid sodium salt 95%

Supplier: Thermo Fisher Scientific

Synonyms: Sodium hyaluronate, Hyaluronic acid sodium, HA sodium salt

251770250
ACRO251770250EA 0 SGD
ACRO251770250
Hyaluronic acid sodium salt 95%
Hyaluronic acid sodium salt

CAS No.: 9067-32-7

Formula: (C₁₄H₂₁NaNO₁₁)n
Storage Temperature: Refrigerator
MDL Number: MFCD00875848
CAS Number: 9067-32-7

Order Now

Specification Test Results

Appearance White to cream Powder
Infrared spectrum Conforms
Assay ≥94 % (On dry substance)
Loss on drying ≤10 % (500 mg, 105°C, 6 h)
Protein ≤0.2 %
Molecular weight 1.5 to 2.2 million Da
pH 6.0 to 8.0 (0.5 % in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR