Glutaraldehyde 8% in aqueous solution

Supplier: Thermo Fisher Scientific
Danger

410971000
ACRO410971000EA 0 SGD
ACRO410971000
Glutaraldehyde 8% in aqueous solution
Glutaraldehyde
Formula: C₅H₈O₂
Storage Temperature: Refrigerator
MDL Number: MFCD00007025
CAS Number: 111-30-8
EINECS: 203-856-5
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear solution
Color scale ≤50 APHA
Specific gravity (20/20°C) 0.918 to 1.123
Titration with NaOH 7 to 9 % v/v

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR