Lecithin (from soy bean), granular

Supplier: Thermo Fisher Scientific

413102500
ACRO413102500EA 0 SGD
ACRO413102500
Lecithin (from soy bean), granular
Lecithin
Density: 1,0305 g/cm³ (20 °C)
MDL Number: MFCD00147406
CAS Number: 8002-43-5
EINECS: 232-307-2
Merck Index: 12,05452

Order Now

Specification Test Results

Appearance Yellow to tan Granules
Infrared spectrum Conforms
Assay ≥97 % (Phospholipids) as Acetone insolubles
Water ≤1.1 % (K.F.)
Insoluble matter ≤0.02 % (in hexane)
Acid value ≤35 mg KOH/g
Peroxide value ≤4 meq O2/kg
UV at 280 nm A: ≤0.1
Residue after ign. (sulfated) ≤0.3 %
Specific optical rotation (after 24 h)
Heavy metals (as Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR