Polyvinylsulphuric acid potassium salt, M.W. ∼ 175,000

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Polyvinyl sulphate potassium salt

418140250
ACRO418140250EA 0 SGD
ACRO418140250
Polyvinylsulphuric acid potassium salt, M.W. ∼ 175,000
Polyvinylsulphuric acid potassium salt

CAS No.: 26837-42-3

Formula: (C₂H₃KO₄S)n
Storage Temperature: Refrigerator
MDL Number: MFCD00081923
CAS Number: 26837-42-3

Order Now

Specification Test Results

Appearance Off-white to beige to light brown Crystalline powder
Infrared spectrum Conforms
Assay ≥21.2 % (K)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR