Sodium hypochlorite 4-6% available chlorine in aqueous solution

Supplier: Thermo Fisher Scientific
Danger

419552500
ACRO419552500EA 0 SGD
ACRO419552500
Sodium hypochlorite 4-6% available chlorine in aqueous solution
Sodium hypochlorite
Formula: ClNaO
Boiling Pt: 111 °C (1013 hPa)
Melting Pt: –30…–20 °C
Density: 1,083 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD00011120
CAS Number: 7681-52-9
EINECS: 231-668-3
UN: 1791
ADR: 8,III

Order Now

Specification Test Results

Appearance (Color) Clear yellow
Appearance (Form) Solution
Titration Iodometric 4.5 to 5.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR