Ethylene glycol, anhydrous 99.8%, AcroSeal®

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Ethane-1,2-diol, 1,2-Dihydroxy ethane, 1,2-Ethanediol, Glycol

433810010
ACRO433810010EA 0 SGD
ACRO433810010
Ethylene glycol, anhydrous 99.8%, AcroSeal®
Ethylene glycol
Formula: HOCH₂CH₂OH
Boiling Pt: 196…198 °C (760 mmHg)
Melting Pt: –13 °C
Density: 1,113 g/cm³
Flash Pt: 116 °C
Storage Temperature: Ambient
MDL Number: MFCD00002885
CAS Number: 107-21-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4d1c97392851ba184f445f6ee912f84631f98d9&vid=2002300f174929a030d66d9f1394eb6bee89b7dc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless Viscous liquid
Infrared spectrum Conforms
Water ≤0.01 %
GC ≥99.75 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR