Precision balances, Highland® series
Supplier: Adam Equipment
HCB123
ADAEHCB123EA 0 SGD
ADAEHCB123
Precision balances, Highland® series
Balances and Scales
The Highland® portable precision balances offers advanced features in a compact, durable balance that stacks for easy storage.
  • Vivid, backlit LCD easily visible in any lighting conditions
  • External calibration allows for verification and adjustment with weights
  • Security lock down
  • Internal rechargeable battery
  • Internal calibration eliminates need for external weight set
  • Dual tare keys operate with either the left or right hand
  • Audible overload alarm warns when capacity is exceeded
The balance has multiple weighing applications including parts counting,More Product Information