Moisture balances, PMB series
Supplier: Adam Equipment
PMB53
ADAEPMB53EA 0 SGD
ADAEPMB53
Moisture balances, PMB series
Analysers Moisture Analysers
The PMB moisture balances offer fast response times and easy-to-use functionality. The automatic test setting function allows multiple tests to be run quickly. The built-in memory permits program storage.
  • Full range tare, zero tracking, capacity tracker
  • Multi-lingual backlit LCD with dual display and colour coded sealed keypad
  • Calibration facilities for temperature (**) and weight
  • RS232 and USB interfaces, plus USB for memory card
  • GLP printouts, date and time function
Other functions include: three settings for heating sample (fast ramp up, step andMore Product Information