Ampoule Filling Jig
Supplier: ADELPHI MANUFACTURING
3510090
ADLM3510090EA 0 SGD
ADLM3510090
Ampoule Filling Jig
Pumps Filling Pumps
A useful hand-operated accessory for use with the Adelphi Accuramatic peristaltic pump (purchased separately). To ensure clean filling of small necked containers such as ampoules, without wetting the wall where it is to be sealed.
  • Reduces waste, increases profitability
  • Simple and intuitive to use
  • Delivery within days
  • Ensures clean filling, without wetting the ampoule wall where it is to be sealed
  • Exceptional filling accuracy of up to ±0,5% reduces product waste and inaccuracies
  • Quick and efficient filling, up to 36 ml per second
  • Hygienic, durable metal
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable