Twin Jet Ampoule Sealer
Supplier: ADELPHI MANUFACTURING
6002100
ADLM6002100EA 0 SGD
ADLM6002100
Twin Jet Ampoule Sealer
Heat Sealers Tube Sealers
A manual solution for consistently high quality ampoule sealing, up to 120/h.
  • Simple and intuitive to use
  • Quick and efficient manual sealing, up to 120/h
  • Sealing open or closed-type ampoules, 1 to 50 ml
  • Supplied with either the air compressor pump, or the compressed air regulator and pipe
  • Gas: Natural gas supply, or bottled butane or propane gas at 0,5 bar
  • Air: Compressed air at 1 bar (optional compressor can be supplied)
Operation – The ampoule is placed and held on the adjustable platform and turned while manually drawing off the ampoule top withMore Product Information