Apollo I liquid viewer, with magnification
Supplier: ADELPHI MANUFACTURING
6031301
ADLM6031301EA 0 SGD
ADLM6031301
Apollo I liquid viewer, with magnification
Lights Light Boxes
Designed for pharmaceutical and laboratory environments, the Apollo I liquid viewer has a polarised inspection light for the detection of particulate matter in transparent liquids contained in ampoules, vials and bottles.
  • Simple detection of unwanted particulate contamination in injections and intravenous infusions
  • Hygienic stainless steel construction
  • LED light source for durable bulb life
  • Intuitive assembly; get started straight away
  • Adjustable height and angle reduce operator fatigue
  • With helpful 1,7× magnifier
Adelphi Manufacturing also offer theMore Product Information