IQ/OQ Documentation for the OC Ampoule Opener and Closer
Supplier: ADELPHI MANUFACTURING
6035000-IQOQ
ADLM6035000-IQOQEA 0 SGD
ADLM6035000-IQOQ
IQ/OQ Documentation for the OC Ampoule Opener and Closer
Services
IQ/OQ documentation for the Adelphi Manufacturing OC ampoule opener and closer.
  • Installation Qualification and Operational Qualification documentation
  • The OC machine itself enables opening and sealing of closed-type ampoules
  • Although specially designed to handle closed-type ampoules, the OC will also handle open-type ampoules after washing
Please note that this listing only refers to the IQ/OQ Documentation, which accompanies the Adelphi Manufacturing OC ampoule opener and closer.
Installation Qualification: Performed to ensure that all key aspectsMore Product Information