OC ampoule sealer and opener, for open and closed type ampoules
Supplier: ADELPHI MANUFACTURING
6035001
ADLM6035001EA 0 SGD
ADLM6035001
OC ampoule sealer and opener, for open and closed type ampoules
Ampoule Openers
The Adelphi Manufacturing OC ampoule opener and sealer is designed to meet the requirements of users who process closed-type ampoules, obviating the need for washing. Suitable for ampoule sizes 1 - 50 ml, this fast, efficient and reliable ampoule sealer gives exceptional output of up to 600 per hour.
  • Ideal for universities
  • Quick and efficient
  • For use with natural gas or propane at 0.5 bar
Operation (Opening): The ampoules are placed into a manually operated spring loaded chuck and a flame is brought to bear on the top of the ampoule. As the flameMore Product Information