Manual Bottle Cap Press
Supplier: ADELPHI MANUFACTURING
SP013-01
ADLMSP013-01EA 0 SGD
ADLMSP013-01
Manual Bottle Cap Press
Crimpers, Cappers and Decappers
This benchtop, hand-operated bottle cap press provides an output up to 1000/hour.
  • Enables easy fixing of press-on lids and caps to glass or plastic bottles
  • Spring-loaded machine
  • Operational weight: 10 kg
Adjustable height and depth allow the device to accommodate a wide range of container heights and cap diameters. It is suitable for use with bottles up to 100 mm in diameter, and up to 240 mm in height.
Adelphi Manufacturing also offers the Pneumatic cap press, with powered operation to reduce operator strain and fatigue.