Semi-Automatic Pneumatic Cap Press
Supplier: ADELPHI MANUFACTURING
SP065-0
ADLMSP065-0EA 0 SGD
ADLMSP065-0
Semi-Automatic Pneumatic Cap Press
Crimpers, Cappers and Decappers
Simple and intuitive fixing of press-on lids and caps to glass or plastic bottles.
  • Quick, efficient, and adaptable
  • Simple and intuitive to use
  • Approximate output of 6 to 10 containers per minute
  • Container diameters from 22 to 85 mm
  • Maximum container height 240 mm
  • Adjustable height and depth to accommodate a wide range of containers
  • Hygienic, durable anodised aluminium and stainless steel construction
Bespoke press pads to suit shaped caps can be made to suit your samples.