Absorbent pads, presterilised
Supplier: Advantech
B200G047A
ADVTB200G047AEA 0 SGD
ADVTB200G047A
Absorbent pads, presterilised
Wipes Cleaning Kits and Pads
Pads are useful for absorbing and retaining media for culturing.
Pads will fit into a standard petri dish, and will absorb 1,8 to 2,2 mL of fluid.
Packaging: Convenient presterilised individual packages.