Platinum ≥99.9% (metals basis), foil, Thickness: 0.25 mm (0.01 in)

Supplier: Thermo Fisher Scientific

Synonyms: Platinium

00861.FF 00861.FI
ALFA00861.FFEA 0 SGD
ALFA00861.FF ALFA00861.FI
Platinum ≥99.9% (metals basis), foil, Thickness: 0.25 mm (0.01 in)
Platinum

Purity: 99.9% CAS No.: 7440-06-4 Physical Form: ≈3.4g/25x25mm Odour: Odourless MDL No.: MFCD00011179

Formula: Pt
Boiling Pt: 3827 °C (1013 hPa)
Melting Pt: 1769 °C
Density: 21,447 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS Number: 7440-06-4
EINECS: 231-116-1
Merck Index: 12,07689

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR